Anti-Glutamic Pyruvic Transaminase Rabbit polyclonal antibody
Catalog # USBIG4010-01A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glutamic Pyruvic Transaminase
- Antigen symbol:GPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2, Dextran.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by solid phase affinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Glutamic Pyruvic Transaminase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: