Anti-Glutamate Decarboxylase 65 Mouse monoclonal antibody [clone: 3H23]
Catalog # USBIG4000-01C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glutamate decarboxylase 65
- Antigen symbol:GAD65
- Clonality:Monoclonal
- Clone:3H23
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:10000-1:100000; Western Blot: 1-10 ?g/ml
Type: Primary
Antigen: Glutamate Decarboxylase 65
Clonality: monoclonal
Clone: 3H23
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat