Order Entry
Netherlands
ContactUsLinkComponent
Anti-Fibroblast Growth Factor-21 Mouse monoclonal antibody [clone: 9H123]
Anti-Fibroblast Growth Factor-21 Mouse monoclonal antibody [clone: 9H123]
Catalog # USBIF4212-93E
Supplier:  US Biological
Anti-Fibroblast Growth Factor-21 Mouse monoclonal antibody [clone: 9H123]
Catalog # USBIF4212-93E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast Growth Factor-21
  • Clonality:
    Monoclonal
  • Clone:
    9H123
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Form:
    lyophilized powder
  • Amino acid number:
    29 to 209
  • Storage buffer:
    PBS, 5% trehalose.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Flow Cytometry: 25 ?g/ml

Type: Primary
Antigen: Fibroblast Growth Factor-21
Clonality: monoclonal
Clone: 9H123
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human