Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-Factor X, Immuno-Adsorbed Sheep Polyclonal Antibody
  USBIF0018-11J
Anti-Factor X, Immuno-Adsorbed Sheep Polyclonal Antibody
  USBIF0018-11J

 

  • Antibody type:
    Primary
  • Antigen name:
    Factor X, Immuno-Adsorbed
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Sheep
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: ELISA: 10 ?g/ml; Western Blot: 10 ?g/ml

Type: Primary
Antigen: Factor X, Immuno-Adsorbed
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Rat