Anti-CRACC Rat Monoclonal Antibody [clone: 9H101]
USBIC7906-72B
- Antibody type:Primary
- Antigen symbol:CRACC
- Clonality:Monoclonal
- Clone:9H101
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:22 to 224
- Storage buffer:PBSwith 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 2?g/ml
Type: Primary
Antigen: CRACC
Clonality: Monoclonal
Clone: 9H101
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse