Order Entry
Netherlands
ContactUsLinkComponent
Anti-Chaperonin 10 Mouse Monoclonal Antibody [clone: 4E44]
Anti-Chaperonin 10 Mouse Monoclonal Antibody [clone: 4E44]
Catalog # USBIC3470A-100
Supplier:  US Biological
Anti-Chaperonin 10 Mouse Monoclonal Antibody [clone: 4E44]
Catalog # USBIC3470A-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chaperonin 10
  • Antigen symbol:
    CPN10
  • Clonality:
    Monoclonal
  • Clone:
    4E44
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Chaperonin 10
Clonality: Monoclonal
Clone: 4E+44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: