Anti-CDw17 Mouse monoclonal antibody [clone: MEM-74]
USBIC2584-01B
- Antibody type:Primary
- Antigen name:CDw17 antigen
- Antigen symbol:CDw17
- Clonality:Monoclonal
- Clone:MEM-74
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CDw17
Clonality: Monoclonal
Clone: MEM-74
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human