Order Entry
Netherlands
ContactUsLinkComponent
Anti-PTPRC Mouse Monoclonal Antibody [clone: MEM-28]
Anti-PTPRC Mouse Monoclonal Antibody [clone: MEM-28]
Catalog # USBIC2400-21D
Supplier:  US Biological
Anti-PTPRC Mouse Monoclonal Antibody [clone: MEM-28]
Catalog # USBIC2400-21D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein tyrosine phosphatase receptor type C
  • Antigen symbol:
    PTPRC
  • Clonality:
    Monoclonal
  • Clone:
    MEM-28
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 15mM sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: Monoclonal
Clone: MEM-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human