Anti-ITGA2B Mouse Monoclonal Antibody [clone: CLB-48]
USBIC2394-02M
- Antibody type:Primary
- Antigen name:Integrin Alpha 2 Beta
- Antigen symbol:ITGA2B
- Clonality:Monoclonal
- Clone:CLB-48
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:20mMTRIS, 150mM sodium chloride, pH 8.0, 10mg BSA, 0,001% Merthiolate.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation and ion exchange chromatography
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: Monoclonal
Clone: CLB-48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human