Anti-CD16,32 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 93]
Catalog # USBIC2268-01G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD16/CD32
- Clonality:Monoclonal
- Clone:93
- Conjugation:PE (Phycoerythrin)/Cy7®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CD16,32
Clonality: Monoclonal
Clone: 93
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse