Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-ITGAL Mouse Monoclonal Antibody [clone: MEM-83]
  USBIC2262-02T
Anti-ITGAL Mouse Monoclonal Antibody [clone: MEM-83]
  USBIC2262-02T

 

  • Antibody type:
    Primary
  • Antigen name:
    Integrin, Alpha L (Antigen CD11A (P180), Lymphocyte Function-Associated Antigen 1, AlphaPolypeptide)
  • Antigen symbol:
    ITGAL
  • Clonality:
    Monoclonal
  • Clone:
    MEM-83
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality: Monoclonal
Clone: MEM-83
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human