Anti-Caldesmon Rabbit Monoclonal Antibody [clone: E-89]
USBIC0132-14Q
- Antibody type:Primary
- Antigen name:Caldesmon
- Clonality:Monoclonal
- Clone:E-89
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Format:Supernatant
- Form:liquid
- Modification:phospho-Ser789
- Storage buffer:PBS, pH 7,4, protebase, sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Recommended dilution: Immunochemistry: 1:25-1:100
Type: Primary
Antigen: Caldesmon
Clonality: Monoclonal
Clone: E-89
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human