Anti-Calcium-Sensing Receptor CASR Rabbit Polyclonal Antibody
Catalog # USBIC0117-06B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Calcium-Sensing Receptor CASR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 15 ?g/ml
Type: Primary
Antigen: Calcium-Sensing Receptor CASR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Rat