Order Entry
Netherlands
ContactUsLinkComponent
Anti-BNP Mouse monoclonal antibody unconjugated [clone: 21-46]
Anti-BNP Mouse monoclonal antibody unconjugated [clone: 21-46]
Catalog # USBIB2702-33D-100
Supplier:  US Biological
Anti-BNP Mouse monoclonal antibody unconjugated [clone: 21-46]
Catalog # USBIB2702-33D-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Brain Natriuretic Peptide
  • Antigen symbol:
    BNP
  • Clonality:
    Monoclonal
  • Clone:
    21-46
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    lyophilized powder
  • Amino acid number:
    1 to 32
  • Storage buffer:
    PBS, pH 7,2
  • Sequence:
    SPKMVQGSGCFGRKMDRISSSSGLGCKVLRRH
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: BNP (Brain Natriuretic Peptide)
Clonality: monoclonal
Clone: 21-46
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human