Order Entry
Netherlands
ContactUsLinkComponent
Anti-Brain Natriuretic Peptide 32 Rabbit polyclonal antibody unconjugated
Anti-Brain Natriuretic Peptide 32 Rabbit polyclonal antibody unconjugated
Catalog # USBIB2702-14H
Supplier:  US Biological
Anti-Brain Natriuretic Peptide 32 Rabbit polyclonal antibody unconjugated
Catalog # USBIB2702-14H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Brain Natriuretic Peptide 32
  • Antigen symbol:
    BNP32
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    400 µg
  • Pk:
    400 µG

Specifications

About this item

Recommended dilution: ELISA: 1:1600

Type: Primary
Antigen: Brain Natriuretic Peptide 32
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat