Anti-PRKAG2 Rabbit Polyclonal Antibody
Catalog # USBIA1475-08
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-Activated Non-Catalytic Subunit Gamma 2
- Antigen symbol:PRKAG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Concentration:~0,05 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: PRKAG2 (protein kinase, AMP-activated, gamma 2 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Monkey, Mouse, Rat