Anti-PDCD6IP Mouse Monoclonal Antibody [clone: 10B133]
Catalog # USBIA1355-64A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Programmed Cell Death 6 Interacting Protein
- Antigen symbol:PDCD6IP
- Clonality:Monoclonal
- Clone:10B133
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Xenopus laevis,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PDCD6IP (programmed cell death 6 interacting protein)
Clonality: monoclonal
Clone: 10B133
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Xenopus laevis