Order Entry
Netherlands
ContactUsLinkComponent
Anti-14-3-3 tau Mouse Monoclonal Antibody [clone: AT1A1]
Anti-14-3-3 tau Mouse Monoclonal Antibody [clone: AT1A1]
Catalog # USBI0014-20G
Supplier:  US Biological
Anti-14-3-3 tau Mouse Monoclonal Antibody [clone: AT1A1]
Catalog # USBI0014-20G
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 tau
  • Clonality:
    Monoclonal
  • Clone:
    AT1A1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    1 to 245
  • Storage buffer:
    PBS, pH 7,4, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: 14-3-3 tau (14-3-3 tau)
Clonality: Monoclonal
Clone: AT1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human