Anti-MYD88 Rabbit Polyclonal Antibody
USBI168380
- Antibody type:Primary
- Antigen name:Myeloid differentiation primary response 88
- Antigen symbol:MYD88
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:0,02M potassium phosphate, 0,15M sodium chloride, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:2000; Immunochemistry: 1:300-1:1000
Type: Primary
Antigen: MYD88 (myeloid differentiation primary response 88)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse