Anti-IFI30 Mouse Monoclonal Antibody [clone: 844623]
Catalog # USBI145490
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interferon-induced Protein 30
- Antigen symbol:IFI30
- Clonality:Monoclonal
- Clone:844623
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: IFI30 (interferon, gamma-inducible protein 30)
Clonality: monoclonal
Clone: 844623
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human