Anti-MAPT Mouse Monoclonal Antibody [clone: TAU-93]
Catalog # USBI145248
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:microtubule-associated Protein tau
- Antigen symbol:MAPT
- Clonality:Monoclonal
- Clone:TAU-93
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Bovine
- Western blot:Yes
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone: TAU-93
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human