Order Entry
Netherlands
ContactUsLinkComponent
Anti-N-Cadherin Mouse Monoclonal Antibody [clone: NC-17]
Anti-N-Cadherin Mouse Monoclonal Antibody [clone: NC-17]
  USBI144943
Anti-N-Cadherin Mouse Monoclonal Antibody [clone: NC-17]
  USBI144943

 

  • Antibody type:
    Primary
  • Antigen name:
    N-Cadherin
  • Clonality:
    Monoclonal
  • Clone:
    NC-17
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Rabbit,
    Chicken,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: N-Cadherin
Clonality: monoclonal
Clone: NC-17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rabbit, Rat