Anti-Pyruvate Dehydrogenase Mouse Monoclonal Antibody [clone: 13k110]
USBI143237
- Antibody type:Primary
- Antigen name:Pyruvate Dehydrogenase
- Clonality:Monoclonal
- Clone:13k110
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:0,1M Tris-glycine, pH 7,4, 150mM NaCl, 0,2% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:150
Type: Primary
Antigen: Pyruvate Dehydrogenase
Clonality: monoclonal
Clone: 13k110
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse