Anti-UNC93B1 Rabbit Polyclonal Antibody
Catalog # USBI138479
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:unc-93 homolog B1 (C. elegans)
- Antigen symbol:UNC93B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS.
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 5 ?g/ml
Type: Primary
Antigen: UNC93B1 (unc-93 homolog B1 (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat