Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-BNIP3L/Nix Rabbit Monoclonal Antibody [clone: 13G20]
  USBI137416
undefined
Anti-BNIP3L/Nix Rabbit Monoclonal Antibody [clone: 13G20]
  USBI137416

 

  • Antibody type:
    Primary
  • Antigen symbol:
    BNIP3L
  • Clonality:
    Monoclonal
  • Clone:
    13G20
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    NIX
  • Storage buffer:
    10mMsodium HEPES, pH 7,5, 150mM NaCl, 100ug/ml BSA, 50% glycerol and <0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Immunofluorescence: 1:200; Western Blot: 1:1000; Immunoprecipitation: 1:100

Type: Primary
Antigen: BNIP3L/Nix
Clonality: monoclonal
Clone: 13G20
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat