Order Entry
Netherlands
ContactUsLinkComponent
Anti-Glucose Regulated Protein 78 Mouse Monoclonal Antibody [clone: 6H4-2G7]
Anti-Glucose Regulated Protein 78 Mouse Monoclonal Antibody [clone: 6H4-2G7]
Catalog # USBI136780
Supplier:  US Biological
Anti-Glucose Regulated Protein 78 Mouse Monoclonal Antibody [clone: 6H4-2G7]
Catalog # USBI136780
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glucose Regulated Protein 78
  • Antigen symbol:
    GRP78
  • Clonality:
    Monoclonal
  • Clone:
    6H4-2G7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,09% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Glucose Regulated Protein 78
Clonality: monoclonal
Clone: 6H4-2G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human