Anti-ZDHHC13 Mouse Monoclonal Antibody [clone: 2A4]
Catalog # USBI135533
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:zinc finger, DHHC-type containing 13
- Antigen symbol:ZDHHC13
- Clonality:Monoclonal
- Clone:2A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:181 to 281
- Storage buffer:PBS, pH 7,2.
- Sequence:NGQTPLMLSAHKVIGPEPTGFLLKFNPSLNVVDKIHQNTPLHWAVAAGNVNAVDKLLEAGSSLDIQNVKGETPLDMALQNKNQLIIHMLKTEAKMRANQK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ZDHHC13 (zinc finger, DHHC-type containing 13)
Clonality: monoclonal
Clone: 2A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human