Anti-Syntaxin-1A Mouse Polyclonal Antibody
USBI134144
- Type antilichaam:Primair
- antigeen symbool:STX1A
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Formulier:liquid
- Opslagbuffer:PBS, pH 7,2.
- Sequentie:MKDRTQELRTAKDSDDDDDVAVTVDRDRFMDEFFEQVEEIRGFIDKIAENVEEVKRKHSAILASPNPDEKTKEELEELMSDIKKTANKVRSKLKSIEQSIEQEEGLNRSSADLRIRKTQHSTLSRKFVEVMSEYNATQSDYRERCKGRIQRQLEITGRTTTSEELEDMLESGNPAIFASGIIMDSSISKQALSEIETRHSEIIKLENSIRELHDMFMDMAMLVESQTMWRGPCLTPRRPSSTRARRAGRKS
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Maat:50 µg
- VPE:50 µG
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: Syntaxin-1A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human