Anti-PDHK2 [Pyruvate Dehydrogenase [Lipoamide]] Kinase Isozyme 2, Mitochondrial, Pyruvate Dehydrogenase Kinase Isoform 2, PDH Kinase 2, PDKII, PDK2) Mouse Monoclonal Antibody [clone: 2G1]
Catalog # USBI131079
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:PDHK2 [Pyruvate Dehydrogenase [Lipoamide]] Kinase Isozyme 2, Mitochondrial, Pyruvate Dehydrogenase Kinase Isoform 2, PDH Kinase 2, PDKII, PDK2)
- Clonality:Monoclonal
- Clone:2G1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:PDKII|PDK2|Pyruvate Dehydrogenase Kinase Isoform 2|PDH Kinase 2
- Storage buffer:PBS, pH 7,2.
- Sequence:HIGSIDPNCNVSEVVKDAYDMAKLLCDKYYMASPDLEIQEINAANSKQPIHMVYVPSHLYHMLFELFKNAMRATVESHESSLILPPIKVM
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 0. 8?g/ml
Type: Primary
Antigen: PDHK2 [Pyruvate Dehydrogenase [Lipoamide]] Kinase Isozyme 2, Mitochondrial, Pyruvate Dehydrogenase Kinase Isoform 2, PDH Kinase 2, PDKII, PDK2)
Clonality: monoclonal
Clone: 2G1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human