Anti-HINT1 Mouse Monoclonal Antibody [clone: 2D7]
Catalog # USBI127857
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Histidine triad nucleotide binding protein 1
- Antigen symbol:HINT1
- Clonality:Monoclonal
- Clone:2D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MADEIAKAQVARPGGDTIFGKIIRKEIPAKIIFEDDRCLAFHDISPQAPTHFLVIPKKHISQISVAEDDDESLLGHLMIVGKKCAADLGLNKGYRMVVNEGSDGGQSVYHVHLHVLGGRQMHWPPG
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: HINT1 (histidine triad nucleotide binding protein 1)
Clonality: monoclonal
Clone: 2D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human