Anti-ERMAP Mouse Monoclonal Antibody [clone: 6F8]
Catalog # USBI126443
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:erythroblast membrane-associated Protein (Scianna blood group)
- Antigen symbol:ERMAP
- Clonality:Monoclonal
- Clone:6F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:376 to 476
- Storage buffer:PBS, pH 7,2.
- Sequence:NKSHIFTFTHNFSGPLRPFFEPCLHDGGKNTAPLVICSELHKSEESIVPRPEGKGHANGDVSLKVNSSLLPPKAPELKDIILSLPPDLGPALQELKAPSF
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ERMAP (erythroblast membrane-associated protein (Scianna blood group))
Clonality: monoclonal
Clone: 6F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human