Anti-ATF7 Mouse Polyclonal Antibody
50 µg
Catalog # USBI123673
Supplier: US Biological
Jump to:
Product Details & Documents
Type: Primary
Antigen: ATF7 (activating transcription factor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Form:liquid
- Amino acid number:1 to 483
- Isotype:IgG
- Storage temperature:Cool
- Host:Mouse
- Sequence:MGDDRPFVCNAPGCGQRFTNEDHLAVHKHKHEMTLKFGPARTDSVIIADQTPTPTRFLKNCEEVGLFNELASSFEHEFKKAADEDEKKAAAGPLDMSLPSTPDIKIKEEEPVEVDSSPPDSPASSPCSPPLKEKEVTPKPVLISTPTPTIVRPGSLPLHLGYDPLHPTLPSPTSVITQAPPSNRQMGSPTGSLPLVMHLANGQTMPVLPGPPVQMPSVISLARPVSMVPNIPGIPGPPVNSSGSISPSGHPIPSEAKMRLKATLTHQVSSINGGCGMVVGTASTMVTARPEQSQILIQHPDAPSPAQPQVSPAQPTPSTGGRRRRTVDEDPDERRQRFLERNRAAASRCRQKRKLWVSSLEKKAEELTSQNIQLSNEVTLLRNEVAQLKQLLLAHKDCPVTALQKKTQGYLESPKESSEPTGSPAPVIQHSSATAPSNGLSVRSAAEAVATSVLTQMASQRTELSMPIQSHVIMTPQSQSAGR
- Shipping temperature:Ice
- Antigen name:Activating transcription factor 7
- Cat. No.:USBI123673
- Reactivity:Human
- Conjugation:Unconjugated
- Antigen symbol:ATF7
- Storage buffer:PBS, pH 7,2.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



