Anti-ASB10 Mouse Monoclonal Antibody [clone: 1F3]
Catalog # USBI123595
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ankyrin repeat and SOCS box containing 10
- Antigen symbol:ASB10
- Clonality:Monoclonal
- Clone:1F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:48 to 153
- Storage buffer:PBS, pH 7,2.
- Sequence:MSWSPEECKGQGEPLDDRHPLCARLVEKPSRGSEEHLKSGPGPIVTRTASGPALAFWQAVLAGDVGCVSRILADSSTGLAPDSVFDTSDPERWRDFRFNIRALRLW
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ASB10 (ankyrin repeat and SOCS box containing 10)
Clonality: monoclonal
Clone: 1F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat