Anti-GIT2 Rabbit Polyclonal Antibody
MEDMMM-0129
: MédiMabs
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor Kinase Interacting ArfGAP 2
- Antigen symbol:GIT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: GIT2 (G protein-coupled receptor kinase interacting ArfGAP 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human