Anti-FGF-21 Mouse Monoclonal Antibody [clone: FG204-3]
Catalog # USBI044798
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:FGF-21
- Clonality:Monoclonal
- Clone:FG204-3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Western blot:Yes
- Form:liquid
- Storage buffer:0,2um-filtered PBS, pH 7,4
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FGF-21
Clonality: Monoclonal
Clone: FG204-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: