Anti-SLC7A6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
USBI043960-HRP
- Type antilichaam:Primair
- antigeen symbool:Solute carrier family 7, member 6
- Clonality:Polyklonaal
- Conjugatie:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flowcytometrie:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitoop:C-terminal
- Formulier:liquid
- Opslagbuffer:PBS, pH 7,2
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Maat:200 µl
- VPE:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Type: Primary
Antigen: SLC7A6 (solute carrier family 7 (amino acid transporter light chain, y+L system), member 6)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human