Anti-SUMF1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBI042467-AP
Supplier: US Biological
Specifications
- Type antilichaam:Primair
- Clonality:Polyklonaal
- Conjugatie:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flowcytometrie:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitoop:C-terminal
- Formulier:liquid
- Aminozuurgetal:310 to 339
- Opslagbuffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Maat:200 µl
- VPE:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: SUMF1 (sulfatase modifying factor 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human