Anti-PIK3R2 Rabbit Polyclonal Antibody (Biotin)
USBI040051-BIOTIN
- Antibody type:Primary
- Antigen name:Phosphoinositide-3-kinase, regulatory subunit 2 (beta)
- Antigen symbol:PIK3R2
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:10-50
Type: Primary
Antigen: PIK3R2 (phosphoinositide-3-kinase, regulatory subunit 2 (beta))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human