Anti-KCNA1 Rabbit Polyclonal Antibody
Catalog # USBI037292
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia)
- Antigen symbol:KCNA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Amino acid number:171 to 201
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Type: Primary
Antigen: KCNA1 (potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human