Anti-TRAF2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 214CT16.3.4]
USBI031306-APC
- Antibody type:Primary
- Antigen name:TNF receptor-associated factor 2
- Antigen symbol:TRAF2
- Clonality:Monoclonal
- Clone:214CT16.3.4
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1000; Immunofluorescence: 1:10-50
Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: Monoclonal
Clone: 214CT16.3.4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human