Order Entry
Netherlands
ContactUsLinkComponent
Anti-MMP2 Mouse Monoclonal Antibody [clone: 6E3F8]
Anti-MMP2 Mouse Monoclonal Antibody [clone: 6E3F8]
Catalogus # USBI031129
Supplier:  US Biological
undefined
Anti-MMP2 Mouse Monoclonal Antibody [clone: 6E3F8]
Catalogus # USBI031129
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)
  • Antigen symbol:
    MMP2
  • Clonality:
    Monoclonal
  • Clone:
    6E3F8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:200-1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: Monoclonal
Clone: 6E3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human