Order Entry
Netherlands
ContactUsLinkComponent
Anti-CLIC5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 16CT28.16.11]
Anti-CLIC5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 16CT28.16.11]
  USBI030878-APC
Anti-CLIC5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 16CT28.16.11]
  USBI030878-APC

 

  • Antibody type:
    Primary
  • Antigen name:
    chloride intracellular channel 5
  • Antigen symbol:
    CLIC5
  • Clonality:
    Monoclonal
  • Clone:
    16CT28.16.11
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG3 kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: CLIC5 (chloride intracellular channel 5)
Clonality: Monoclonal
Clone: 16CT28.16.11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human, Mouse