Anti-CLIC5 Mouse Monoclonal Antibody [clone: 16CT28.16.11]
Catalog # USBI030878
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:chloride intracellular channel 5
- Antigen symbol:CLIC5
- Clonality:Monoclonal
- Clone:16CT28.16.11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:10-50
Type: Primary
Antigen: CLIC5 (chloride intracellular channel 5)
Clonality: Monoclonal
Clone: 16CT28.16.11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human, Mouse