Anti-ATG5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 22CT1.11.4.4]
Catalog # USBI030779-PE
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Autophagy related 87
- Antigen symbol:ATG87
- Clonality:Monoclonal
- Clone:22CT1.11.4.4
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:APG5
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:50-100
Do not freeze! Light sensitive
Type: Primary
Antigen: ATG5 (autophagy related 5)
Clonality: Monoclonal
Clone: 22CT1.11.4.4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human