Anti-SMN1 Mouse Monoclonal Antibody [clone: 13B714]
USBI030636
- Antibody type:Primary
- Antigen name:survival of motor neuron 1, telomeric
- Antigen symbol:Survival of Motor Neuron 1
- Clonality:Monoclonal
- Clone:13B714
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western blot:Yes
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: SMN1 (survival of motor neuron 1, telomeric)
Clonality: Monoclonal
Clone: 13B714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey