Anti-MST1R Mouse Monoclonal Antibody [clone: 13B680]
USBI030602
- Antibody type:Primary
- Antigen name:macrophage stimulating 1 receptor (c-met-related tyrosine kinase)
- Antigen symbol:MST1R
- Clonality:Monoclonal
- Clone:13B680
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Amino acid number:210 to 320
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: MST1R (macrophage stimulating 1 receptor (c-met-related tyrosine kinase))
Clonality: Monoclonal
Clone: 13B680
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human