Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-MAP Kinase p44/42 Mouse Monoclonal Antibody [clone: 13B490]
  USBI030412
Anti-MAP Kinase p44/42 Mouse Monoclonal Antibody [clone: 13B490]
  USBI030412

 

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase p44/42
  • Clonality:
    Monoclonal
  • Clone:
    13B490
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Format:
    ascites
  • Storage buffer:
    0,03% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: Monoclonal
Clone: 13B490
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse