Order Entry
Netherlands
ContactUsLinkComponent
Anti-Bone Morphogenic Protein 14 Rabbit Polyclonal Antibody
Anti-Bone Morphogenic Protein 14 Rabbit Polyclonal Antibody
Catalog # USBI029739
Supplier:  US Biological
Anti-Bone Morphogenic Protein 14 Rabbit Polyclonal Antibody
Catalog # USBI029739
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Bone Morphogenic Protein 14
  • Antigen symbol:
    BMP14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 1% BSA, 0,02% thimerosal, 30%glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: Bone Morphogenic Protein 14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human