Anti-SPAK+OSR1 Rabbit Polyclonal Antibody
Catalog # USBI029614
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:SPAK+OSR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser373, phospho-Ser325
- Storage buffer:0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: SPAK+OSR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse