Anti-Fcgr3 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 93]
ROCK200-508-N80
- Antibody type:Primary
- Antigen name:Fc fragment of IgG, low affinity II/III
- Antigen symbol:FCGR2/3
- Clonality:Monoclonal
- Clone:93
- Conjugation:RPE (R-Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:Sufficient to run approximately 100 tests
- Shipping temperature:Ice
- Immunogen:Anti-CD16/32 Antibody (Monoclonal) was produced by repeated immunizations with sorted pre-B cells.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:200 µg
- Pk:200 µG
Anti-CD16/32 is useful for Immunoprecipitation and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!
Type: Primary
Antigen: Fcgr3
Clonality: Monoclonal
Clone: 93
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse